Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRTC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584611
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584611 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584611 Supplier Invitrogen™ Supplier No. PA584611
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSLTNFFPDV GFDQQSMRPG PAFPQQVPLV QQGSRELQDS FHLRPSPYSN CGSLPNTILP EDSSTS Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 74%.

The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTC3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Crtc3
Gene Accession No. Q6UUV7
Gene Alias 2610312F20Rik; 6332415K15Rik; AI429792; BC011210; CREB regulated transcription coactivator 3; CREB-regulated transcription coactivator 3; Crtc3; RGD1309666; TORC3; TORC-3; transducer of CREB protein 3; transducer of regulated cAMP response element-binding protein (CREB) 3; transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3
Gene Symbols Crtc3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRTC3. Recombinant protein control fragment (Product #RP-105271).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64784
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.