Learn More
Thermo Scientific™ Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Catalog No. p-7220055
Encompass
Click to view available options
Type:
ICP-MS Organic Torch
Get improved plasma stability from the tulip design and easy, push-in insertion of Thermo Scientific™ torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Gain improved plasma stability from the tulip design and easy, push-in Gain improved plasma stability from the tulip design and easy, push-in insertion of fully demountable, single piece, Thermo Scientific™ Torches with connection-free gas supply for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems. All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
TRUSTED_SUSTAINABILITY
Specifications
Type ICP-MS Organic Torch
For Use With (Equipment) iCAP MX and iCAP Q/Qnova Series ICP-MS Systems
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.