Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Torin 1
GREENER_CHOICESDP

Catalog No. 424750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-475-0 50 mg
42-471-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-475-0 Supplier R&D Systems™ Supplier No. 4247/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective mTOR inhibitor

Torin 1 is a potent and selective ATP-competitive mTOR inhibitor (IC50 = 2 - 10 nM for mTORC1 and mTORC2). Torin 1 displays 200-fold selectivity for mTOR over DNA-PK, ATM and hVps34. In HeLa cells, Torin 1 induces autophagy. Torin 1 impairs cell growth and proliferation by suppression of the rapamycin-resistant functions of mTORC1. In human monocytes and myeloid dendritic cells, Torin 1 prevents decreases the anti-inflammatory potency of glucocorticoids. In the human endocrine cell line BON, Torin 1 increases neurotensin secretion and gene expression through MEK/ERK/c-Jun pathway activation.

Chemical Identifiers

CAS 1222998-36-8
Molecular Formula C35H28F3N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 607.62
Purity >98%
Molecular Weight (g/mol) 607.64
Chemical Name or Material Torin 1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.