Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Torpedo californica CHRNA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117457
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117457 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117457 Supplier Invitrogen™ Supplier No. PA5117457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Acetylcholine receptors (AChRs) mediate synaptic transmission at the neuromuscular junction. The channel-linked AChR that mediates rapid, excitatory actions of acetylcholine is called nicotinic AChR (nAChR) because it can be activated by nicotine. The non-channel linked AChR that medicates the slow actions of acetylcholine, which can be either inhibitory or excitatory, is called muscarinic AChR (mAChR) because it can be activated by muscarine.The nAChRs fall into two subclasses, the muscle-type and the neuronal-type. The muscle-type receptor is a pentameric glycoprotein of five membrane spanning subunits, (2 alpha 1's, beta 1, gamma or epsilon and delta) which form a ligand gated ion channel. The neuronal nAChR is also a pentamer, but unlike the muscle-type, is made up of only two general types of subunits, alpha and beta of which multiple subtypes have been described (alpha 2 to alpha 8 and beta 2 to beta 4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Torpedo californica CHRNA1
Applications Western Blot
Classification Polyclonal
Concentration 2.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias acetylcholine receptor; Acetylcholine receptor subunit alpha; acetylcholine receptor, nicotinic, alpha 1 (muscle); Achr-1; ACHRA; ACHRD; Acra; AI385656; AI608266; alpha-1 subunit, nicotinic acetylcholine receptor; CHNRA; cholinergic receptor; cholinergic receptor nicotinic alpha 1 subunit; cholinergic receptor nicotinic alpha polypeptide 1; cholinergic receptor, nicotinic alpha 1; cholinergic receptor, nicotinic, alpha 1 (muscle); cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle); CHRNA; CHRNA1; CMS1A; CMS1B; CMS2A; FCCMS; muscle nicotinic acetylcholine receptor; nAChR; NACHRA1; nicotinic acetylcholine receptor alpha subunit; nicotinic acetylcholine receptor, alpha-1 subunit; nicotinic cholinergic receptor alpha 1; SCCMS
Host Species Rabbit
Immunogen Recombinant Torpedo californica Acetylcholine receptor subunit alpha protein (25-234aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fish
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.