Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Torsin 1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102569
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102569 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102569 Supplier Invitrogen™ Supplier No. PA5102569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TOR1A.

The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Torsin 1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Tor1a
Gene Accession No. O14656, Q68G38, Q9ER39
Gene Alias DQ2; dystonia 1; Dystonia 1 protein; Dystonia 1 torsion; dystonia 1, torsion (autosomal dominant; dystonia 1, torsion (autosomal dominant; torsin A); DYT1; LOW QUALITY PROTEIN: torsin-1A; TA; Tor1a; TORA; Torsin 1A; torsin A; torsin ATPase 1; Torsin ATPase-1A; torsin family 1 member A; torsin family 1, member A; torsin family 1, member A (torsin A); torsin-1A; torsinA
Gene Symbols Tor1a
Host Species Rabbit
Immunogen A synthesized peptide derived from human TOR1A(Accession O14656), corresponding to amino acid residues G225-K275.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1861, 266606, 30931
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.