Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Torsin 1A Recombinant Rabbit Monoclonal Antibody (4Q2I9)
GREENER_CHOICE

Catalog No. PIMA542926
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542926 Supplier Invitrogen™ Supplier No. MA542926
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: 293T, HepG2, U-87MG, Mouse liver, Mouse brain, Mouse spleen, Rat lung. Immunogen sequence: LKDIEHALSV SVFNNKNSGF WHSSLIDRNL IDYFVPFLPL EYKHLKMCIR VEMQSRGYEI DEDIVSRVAE EMTFFPKEER VFSDKGCKTV FTKLDYYYDD.

The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Torsin 1A
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Q2I9
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Tor1a
Gene Accession No. O14656, Q68G38, Q9ER39
Gene Alias DQ2; dystonia 1; Dystonia 1 protein; Dystonia 1 torsion; dystonia 1, torsion (autosomal dominant; dystonia 1, torsion (autosomal dominant; torsin A); DYT1; LOW QUALITY PROTEIN: torsin-1A; TA; Tor1a; TORA; Torsin 1A; torsin A; torsin ATPase 1; Torsin ATPase-1A; torsin family 1 member A; torsin family 1, member A; torsin family 1, member A (torsin A); torsin-1A; torsinA
Gene Symbols Tor1a
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1861, 266606, 30931
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.