Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Torsin-4A Antibody (SR1822), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | Torsin-4A |
---|---|
Clone | SR1822 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Torsin-4A Monoclonal antibody specifically detects Torsin-4A in Human samples. It is validated for Western BlotSpecifications
Torsin-4A | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
54863 | |
IgG | |
Affinity purified |
SR1822 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C9orf167, chromosome 9 open reading frame 167, FLJ20245, RP13-122B23.4, torsin family 4, member A | |
A synthesized peptide derived from human Torsin-4A (Uniprot #: Q9NXH8) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title