Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tositumomab Recombinant Mouse Monoclonal Antibody
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA541763
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Murine IgG2a lambda monoclonal antibody against CD20 antigen (2 heavy chains of 451 residues, 2 lambda chains of 220 residues). It is produced in an antibiotic-free culture of mammalian cells. It can be covalently linked to Iodine 131 (a radioactive isotope of iodine). Binds to the CD20 antigen which is found on mature B lymphocytes. The antibody binding appears to induce apoptosis, complement-dependent cytotoxicity and cell death through ionizing radiation.
Specifications
Tositumomab | |
Recombinant Monoclonal | |
Unconjugated | |
Bexxar | |
Human MS4A1/CD20. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction