Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tosylmethyl isocyanide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
36635-61-7
C9H9NO2S
195.24
MFCD00000005
CFOAUYCPAUGDFF-UHFFFAOYSA-N
tosylmethyl isocyanide, tosmic, p-toluenesulfonylmethyl isocyanide, 4-toluenesulphonylmethyl isocyanide, 4-toluenesulfonylmethyl isocyanide, tosylmethylisocyanide, 1-isocyanomethylsulfonyl-4-methylbenzene, p-tolylsulfonyl methyl isocyanide, unii-c35fd6olh8, 1-isocyanomethyl sulfonyl-4-methylbenzene
161915
1-(isocyanomethylsulfonyl)-4-methylbenzene
CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]
36635-61-7
C9H9NO2S
195.24
MFCD00000005
CFOAUYCPAUGDFF-UHFFFAOYSA-N
tosylmethyl isocyanide, tosmic, p-toluenesulfonylmethyl isocyanide, 4-toluenesulphonylmethyl isocyanide, 4-toluenesulfonylmethyl isocyanide, tosylmethylisocyanide, 1-isocyanomethylsulfonyl-4-methylbenzene, p-tolylsulfonyl methyl isocyanide, unii-c35fd6olh8, 1-isocyanomethyl sulfonyl-4-methylbenzene
161915
1-(isocyanomethylsulfonyl)-4-methylbenzene
CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]
Infrared Spectrum Authentic
Color Undesignated
Packaging Glass bottle
Melting Point 111.0°C to 113.0°C
Loss on Drying 2% max.
Quantity 5g
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CH3C6H4SO2CH2NC
Fieser 04,514; 05,684; 06,600; 07,377; 08,493; 10,409; 11,539; 12,511; 13,313; 17,20
Merck Index 15, 9715
Solubility Information Solubility in water: insoluble
Formula Weight 195.24
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Tosylmethyl isocyanide
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 253-140-1