Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tosylmethyl isocyanide, 98%

Catalog No. p-3737591
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 36635-61-7 | C9H9NO2S | 195.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36635-61-7
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00000005
InChI Key CFOAUYCPAUGDFF-UHFFFAOYSA-N
Synonym tosylmethyl isocyanide, tosmic, p-toluenesulfonylmethyl isocyanide, 4-toluenesulphonylmethyl isocyanide, 4-toluenesulfonylmethyl isocyanide, tosylmethylisocyanide, 1-isocyanomethylsulfonyl-4-methylbenzene, p-tolylsulfonyl methyl isocyanide, unii-c35fd6olh8, 1-isocyanomethyl sulfonyl-4-methylbenzene
PubChem CID 161915
IUPAC Name 1-(isocyanomethylsulfonyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C[N+]#[C-]

Specifications

Melting Point 111.0°C to 113.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H4SO2CH2NC
Quantity 5 g
Fieser 04,514; 05,684; 06,600; 07,377; 08,493; 10,409; 11,539; 12,511; 13,313; 17,20
Merck Index 15, 9715
Solubility Information Solubility in water: insoluble
Formula Weight 195.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tosylmethyl isocyanide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-140-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.