Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Total BDNF Quantikine ELISA Kit

Catalog No. dbnt00
Click to view available options
Conjugate:
HRP
This item is not returnable. View return policy
This item is not returnable. View return policy

Total BDNF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Total BDNF Quantikine ELISA Kit quantifies human, mouse, rat, porcine, canine BDNF in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (50 ul), platelet-poor edta plasma (10 ul), heparin plasma (50 ul), platelet-poor heparin plasma (10 ul), urine (50 ul), human milk (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 1.35pg/mL
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested.
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Serum, Tissue Lysates, Urine
Specificity Natural and Recombinant free, trk-bound, pro-, and mature BDNF.
For Use With (Application) ELISA
Gene ID (Entrez) 627 (Human); 12064 (Mouse); 24225 (Rat)
Sample Volume Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (50uL),Platelet-poor EDTA Plasma (10uL),Heparin Plasma (50uL),Platelet-poor Heparin Plasma (10uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.