Learn More
Description
Includes
Total bilirubin reagent: Sulfanilic acid 16mM, hydrochloric acid 164mM, dimethyl sulfoxide 4.4M, surfactant; Sodium nitrite reagent: Sodium nitrite 0.6M
- Method: Walters and Gerarde, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 550nm
- Linearity: 20mg/dL
- Expected Values: 0.2 to 1.2mg/dL
For the quantitative determination of total bilirubin in serum.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Total Bilirubin Reagent Kit |
| Formulation | Total Bilirubin reagent: Sulfanilic Acid 16mM, Hydrochloric Acid 164mM, Dimethyl Sulfoxide 4.4M, Surfactants, Sodium Nitrite 0.6M |
| For Use With (Application) | Quantitative determination of total Bilirubin in Serum, in-vitro diagnostic use only |
| Quantity | 1 x 120 mL |
| Wavelength | 550 nm |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.