Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix, Reagents

Catalog No. 01335318
Click to view available options
Quantity:
100 g
100 mL

Catalog No. 01-335-318

Supplier: Reagents Holdings Llc CE255600100A6

Only null left
Add to Cart
Add to Cart

Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix
CAS 7778-80-5, 7778-77-0
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.372
Color Purple
Quantity 100 mL
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.