Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix, Reagents
Click to view available options
Quantity:
100 g
100 mL
Specifications
Specifications
| Chemical Name or Material | Total Hardness Indicator Eriochrome Black T Indicator, Dry Mix |
| CAS | 7778-80-5, 7778-77-0 |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 2.372 |
| Color | Purple |
| Quantity | 100 mL |
| Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction