Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA, Ricca Chemical
SDP

Catalog No. 867032
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

For Fluoride Analysis using Ion Selective Electrodes

Specifications

Chemical Name or Material Buffer Solution, Total Ionic Strength Adjustment Buffer (TISAB II)
Name Note With CDTA
CAS 7732-18-5, 1310-73-2, 64-19-7, 7647-14-5, 13291-61-7
CAS Min % 82.0, 5.87, 4.91, 4.77, 0.33
CAS Max % 85.0, 6.11, 5.12, 4.96, 0.34
Concentration 1X
For Use With (Application) Fluoride Analysis
For Use With (Equipment) Ion Selective Electrode
pH 5.4
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.