Learn More
Total Organic Carbon (TOC), standard solution, Specpure™ 1000μg/mL
Description
Total Organic Carbon (TOC), is used as a standard solution during the measurement of chemical analysis such as detecting contaminants in drinking water and cooling water. It is also used for analysis of water to be used for semiconductor manufacturing and pharmaceutical use.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Chemical Name or Material | Total Organic Carbon |
Name Note | 1000μg/mL |
Odor | Characteristic |
Physical Form | Liquid |
Molecular Formula | KHC8H4O4 in H2O |
MDL Number | MFCD00133992 |
Color | Colorless |
Solubility | Miscible with water |
Quantity | 500 mL |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.