Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Total p38 alpha DuoSet IC ELISA
SDP

Catalog No. DYC8691B5 Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC8691B5 5 Plates
DYC8691B2 2 Plates
DYC8691BE 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC8691B5 Supplier R&D Systems™ Supplier No. DYC8691B5
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Total p38 alpha DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Total p38 alpha DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat p38 alpha.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit (for 5 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 1432
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.