Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell TouchNTuff™ 92-600 Nitrile Gloves

Catalog No. 19010412 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5, 7
7.5, 8
8.5, 9
9.5, 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-010-412 6.5, 7
19-010-413 7.5, 8
19-010-414 8.5, 9
19-010-415 9.5, 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-010-412 Supplier Ansell Supplier No. 585834/DEL
This item is not returnable. View return policy

Ansell™ TouchNTuff™ 92-600 Nitrile Gloves offers superior splash resistance against a wide range of hazardous chemicals.

  • Enhanced chemical splash protection for longer periods
  • Offers superior comfort due to proprietary nitrile formulation
  • Delivers strong grip in wet or dry conditions
  • Highly versatile and suitable for many different uses
  • Silicone free for product protection
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6319, FDA 21 CFR 177-2600
Color Green
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 6.5, 7
Sterility Non-sterile
For Use With (Application) Laboratory analysis, Electronic manufacture, Pharmaceuticals
Glove Cuff Style Beaded
Quantity 100 gloves per dispensers, 10 dispensers per shipper carton, 1000 gloves per shipper carton
Length (Metric) 24 cm
Show More Show Less

*Warning: No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.