Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540307 Supplier Invitrogen™ Supplier No. PA540307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TSQTYNGQSE NNEDYEIPPI TPPNLPEPSL LHLGDHEASY HSLCHGLTPN Sequence homology: Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

TOX2 (TOX High Mobility Group Box Family Member 2) is a Protein Coding gene. Diseases associated with TOX2 include Conduct Disorder. Gene Ontology (GO) annotations related to this gene include chromatin binding and RNA polymerase II transcription factor binding. An important paralog of this gene is TOX.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOX2
Applications Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TOX2
Gene Accession No. Q96NM4
Gene Alias AI851523; AV026525; BI987407; C20orf100; dJ1108D11.2; dJ495O3.1; GCX1; Gcx-1; granulosa cell HMG box 1; granulosa cell HMG box protein 1; granulosa cell HMG-box protein 1; granulosa cell HMG-box protein-1; LOW QUALITY PROTEIN: TOX high mobility group box family member 2; RxHMG1; TOX high mobility group box family member 2; TOX2
Gene Symbols TOX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C20ORF100 (aa 62-111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84969
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.