Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOX3 Mouse anti-Human, Unconjugated, Clone: TOX3/1124, Abnova™

Catalog No. 89337510 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-510 Supplier Abnova Corporation Supplier No. MAB21017
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

TOX3 Monoclonal antibody specifically detects TOX3 in Human samples. It is validated for WB-Ce, IF, Flow Cyt.

Specifications

Antigen TOX3
Applications Western Blot, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone TOX3/1124
Conjugate Unconjugated
Description TOX high mobility group box family member 3
Gene TOX3
Gene Alias CAGF9, TNRC9
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 200-400 of human TOX3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 27324
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.