Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112771
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112771 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112771 Supplier Invitrogen™ Supplier No. PA5112771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOX3 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

TOX3 contains an HMG-box, indicating that it may be involved in bending and unwinding of DNA and alteration of chromatin structure. The C-terminus of the encoded protein is glutamine-rich due to CAG repeats in the coding sequence. A minor allele of this gene has been implicated in an elevated risk of breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOX3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TOX3
Gene Accession No. O15405, Q80W03
Gene Alias 500-9; BC052044; C230068E13; CAG trinucleotide repeat-containing gene F9 protein; CAGF9; Tnrc9; TOX high mobility group box family member 3; TOX3; trinucleotide repeat containing 9; trinucleotide repeat-containing gene 9 protein
Gene Symbols TOX3
Host Species Rabbit
Immunogen Recombinant Human TOX high mobility group box family member 3 protein (8-305AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 244579, 27324
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.