Learn More
Invitrogen™ TOX4 Polyclonal Antibody

Description
Peptide sequence: RCVRSGCENP PIVSKDWDNE YCSNECVVKH CRDVFLAWVA SRNSNTVVFV Sequence homology: Cow: 93%; Dog: 93%; Goat: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 86%; Zebrafish: 77%.
Specifications
Specifications
| Antigen | TOX4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TOX4 |
| Gene Accession No. | O94842 |
| Gene Alias | 5730589K01Rik; A630040M18; AA410149; C14orf92; Epidermal Langerhans cell protein LCP1; KIAA0737; LCP1; MIG7; migration-inducing protein 7; RGD708449; TOX high mobility group box family member 4; TOX4; Unknown (protein for MGC:139878) |
| Gene Symbols | TOX4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.