Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553653 Supplier Invitrogen™ Supplier No. PA553653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQQPPPQKVR INLQQQPPPL QIKSVPLPTL KMQTTLVPPT VESSPERPMN NSPEAHTVEA PSPETICEMI TDVVPEVESP SQMDVELVSG SPVALSPQPR CVRSGCENPP IVSKDWDNEY CSNECVVKHC RDVFLAWVAS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.

KIAA0737 contains 1 HMG box DNA-binding domain and the function remains unknows.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOX4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOX4
Gene Accession No. O94842, Q8BU11, Q99PM1
Gene Alias 5730589K01Rik; A630040M18; AA410149; C14orf92; Epidermal Langerhans cell protein LCP1; KIAA0737; LCP1; MIG7; migration-inducing protein 7; RGD708449; TOX high mobility group box family member 4; TOX4; Unknown (protein for MGC:139878)
Gene Symbols TOX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOX4. Recombinant protein control fragment (Product #RP-88896).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268741, 286990, 9878
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.