Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxaphene (For EPA 500, 600, 8000/; Appendix IX, CLP Methods)

Catalog No. p-220649
Encompass
Click to view available options
Quantity:
1 mL (in Hexane)
1 mL (in Methanol)
Packaging:
Ampule

Toxaphene at 100μg/mL in methanol or hexane

Chemical Identifiers

CAS 8001-35-2

Specifications

Packaging Ampule
Methods For EPA 500, 600, 8000/Appendix IX
Quantity 1 mL (in Hexane)
Chemical Name or Material Toxaphene
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.