Learn More
Invitrogen™ Toxoplasma gondii 38 kDa Monoclonal Antibody (J3D)

Description
This antibody recognizes a ~38 kDa protein of T. gondii. Purity is >90%.
Specifications
Specifications
| Antigen | Toxoplasma gondii 38 kDa |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | J3D |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | t gondii; T. gondii |
| Host Species | Mouse |
| Immunogen | Toxoplasma gondii native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.