Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Toxoplasma gondii P30 Monoclonal Antibody (J26A), FITC
GREENER_CHOICE

Catalog No. PIMA17436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17436 Supplier Invitrogen™ Supplier No. MA17436
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7436 antibody reacts with Toxoplasma gondii in bacterial and infected tissue samples. MA1-7436 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7436 antibody was raised against Whole organism preparation.

Toxoplasma is a crescent shaped sporozoan that lives as intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregnant women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregnancy, can produce blindness or mental retardation in the newborn. Toxoplasma gondii infections in humans are widespread but vary greatly by region. The organism enters the intestinal epithelium and can spread to other host tissues. In healthy individuals the infection is usually asymptomatic. However, for individuals with compromised immune systems, a Toxoplasma infection can cause serious health problems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Toxoplasma gondii P30
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone J26A
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Gene Alias t gondii; T. gondii; T. gondii P30
Host Species Mouse
Immunogen Whole organism preparation.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.