Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxoplasma Gondii P30 Antibody (P30/3), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1066486A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066486A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066486A70 Supplier Novus Biologicals Supplier No. NB10066486AF750

Mouse Monoclonal Antibody

Toxoplasma Gondii P30 Monoclonal specifically detects Toxoplasma Gondii P30 in Protozoa, T. gondii samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Toxoplasma Gondii P30
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone P30/3
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias p30, SAG1, SAG1 protein
Host Species Mouse
Immunogen Purified T. gondii P30 antigen
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for the Toxoplasma gondii P30 membrane protein.
Target Species Protozoa, T. gondii
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.