Learn More
Invitrogen™ Toxoplasma gondii Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA17256
Description
PA1-7256 detects the Toxoplasma gondii. PA1-7256 has been successfully used in immunofluorescence procedures. The PA1-7256 immunogen is RH strain of Toxoplasma gondii. Recommended short term (<6 months) storage is liquid at 2-8ºC. For longer term storage, aliquot and freeze.
Toxoplasma is a crescent shaped sporozoan that lives as an intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregnant women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregnancy, can produce blindness or mental retardation in the newborn.
Specifications
Toxoplasma gondii | |
Polyclonal | |
Unconjugated | |
t gondii; T. gondii | |
RH strain of Toxoplasma gondii. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Protozoa | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.