Learn More
Invitrogen™ Toxoplasma gondii Polyclonal Antibody, FITC

Description
PA1-7253 detects Toxoplasma gondii. PA1-7253 has been successfully used in immunofluorescence procedures. The PA1-7253 immunogen is RH strain of Toxoplasma gondii.
Specifications
Specifications
| Antigen | Toxoplasma gondii |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene Alias | t gondii; T. gondii |
| Host Species | Rabbit |
| Immunogen | RH strain of Toxoplasma gondii. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.