Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxoplasma gondii SAG2 Antibody (1D12B8) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-200080634 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61907 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61907 Supplier Novus Biologicals Supplier No. NBP261907
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Toxoplasma gondii SAG2 Monoclonal specifically detects Toxoplasma gondii SAG2 in T. gondii samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Toxoplasma gondii SAG2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1D12B8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias Toxoplasma gondii surface antigen P22
Gene Symbols SAG2
Host Species Mouse
Immunogen Purified recombinant fragment of Toxoplasma gondii surface antigen 2B SAG2 (AA: 1-130) expressed in E. Coli.
Molecular Weight of Antigen 110 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species T. gondii, Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.