Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toxoplasma Gondii P30 Antibody (P30/3), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066486JF646
Description
Toxoplasma Gondii P30 Monoclonal specifically detects Toxoplasma Gondii P30 in Protozoa, T. gondii samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
ToxoplasmaGondiiP30 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein G purified | |
Primary | |
Protozoa, T. gondii | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
P30/3 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
p30, SAG1, SAG1 protein | |
Purified T. gondii P30 antigen | |
0.1 mL | |
Specific for the Toxoplasma gondii P30 membrane protein. | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction