Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOXR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144481
Description
TOXR Polyclonal Antibody for Western Blot, ELISA
Positive regulation of toxA gene transcription.
Specifications
TOXR | |
Polyclonal | |
Unconjugated | |
toxR | |
regA | |
Rabbit | |
Protein A/G | |
RUO | |
880684 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.68 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P09852 | |
toxR | |
Recombinant Pseudomonas aeruginosa (strain ATCC 15692/DSM 22644/CIP 104116/JCM 14847/LMG 12228/1C/PRS 101/PAO1) TOXR protein. | |
100 μL | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction