Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TP 003
GREENER_CHOICESDP

Catalog No. 441450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-145-0 50 mg
44-141-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-145-0 Supplier Tocris Bioscience™ Supplier No. 4414/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

GABAA partial agonist; acts at benzodiazepine site

GABAA receptor partial agonist that demonstrates affinity for the benzodiazepine site. Potentiates GABAA α3 response with high efficacy. Produces anxiolytic-like effects in rodent elevated plus maze via action at GABAA α2 subtype receptors. Also evokes GABAA α1 mediated sedation and GABAA α3 mediated muscle relaxation in mice.

Chemical Identifiers

CAS 628690-75-5
Molecular Formula C23H16F3N3O

Specifications

Quantity 50 mg
Formula Weight 407.39
Purity >98%
Molecular Weight (g/mol) 407.396
Chemical Name or Material TP 003
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.