Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TP 064
GREENER_CHOICESDP

Catalog No. 600810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-081-0 10 mg
60-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-081-0 Supplier Tocris Bioscience™ Supplier No. 6008/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PRMT 4 inhibitor

TP 064 is a potent and selective PRMT4 inhibitor; inhibits methylation of H3 (1-25) and MED12 (IC50 values are <10 and 43 nM, respectively). Exhibits >100-fold selectivity over other histone methyltransferases and non-epigenetic targets. Inhibits proliferation of certain multiple myeloma cell lines, arresting cells in G1 phase. TP 064 can be used in vivo. TP 064 inhibits the monocyte/macrophage-driven inflammation in ex vivo.

To request the negative control for TP 064, please fill out the TP 064N request form on the SGC website..

Specifications

Chemical Name or Material TP 064
Synonym N-Methyl-N-((2-(1-(2-(methylamino)ethyl)piperidin-4-yl)pyridin-4-yl)methyl)-3-phenoxybenzamide
Quantity 10 mg
Solubility Soluble to 100mM in 1eq. HCl and to 100mM in DMSO
Molecular Formula C28H34N4O2
Formula Weight 458.6
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.