Learn More
Description
A concentration of no more than 2 μM is recommended for use in cellular assays.
Specifications
Specifications
| Chemical Name or Material | N-[(3-(1 H)-Pyrazol-1-yl)propyl)-6-(4-(3-(dimethylamino)propoxy)phenyl)-2-(methylsulfonyl)pyrimidin-4-amine hydrochloride |
| CAS | 2415263-05-5 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Bromodomain Inhibitors |
| Molecular Formula | C22H30N6O<sub>3S.HCl |
| Formula Weight | 2576 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

