Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TP 472N
GREENER_CHOICESDP

Catalog No. 599910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-991-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-991-0 Supplier R&D Systems™ Supplier No. 5999/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Negative control for TP 472

TP 472N is a negative control for TP 472 . Inactive against other bromodomains (>20 μM against BRD9).

Active Analog also available.

Specifications

Chemical Name or Material TP 472N
Synonym 1-(8-(2-(Cyclopropylmethoxy)phenyl)pyrrolo[1,2-a]pyrimidin-6-yl)ethanone
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Product Line Tocris
Molecular Formula C19H18N2O2
Formula Weight 306.36
Physical Form Solid
Color Yellow
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.