Learn More
Description
Mouse monoclonal antibody raised against full length recombinant human TP53.
- High specificity
- High reactivity
Specifications
Specifications
| Antigen | TP53 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | TRP/817 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against full length recombinant human TP53. |
| Dilution | Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determin |
| Formulation | In 10 mM PBS (0.05% BSA and 0.05% azide). |
| Gene Accession No. | P04637 |
| Gene Alias | FLJ92943/LFS1/TRP53/p53 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.