Learn More
Invitrogen™ TP53I11 Polyclonal Antibody

Description
Immunogen sequence: MAAKQPPPLM KKHSQTDLVS RLKTRKILGV GGEDDDGEVH RSKISQVLGN EIKFTIREPL GLRVWQFVSA; Positive Samples: HeLa, Mouse brain; Cellular Location: Membrane, Multi-pass membrane protein.
Specifications
Specifications
| Antigen | TP53I11 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.69 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | TP53I11 |
| Gene Accession No. | B3DMA0, O14683 |
| Gene Alias | p53-induced gene 11 protein; Pig11; TP53I11; transformation related protein 53 inducible protein 11; Trp53i11; tumor protein p53 inducible protein 11; tumor protein p53-inducible protein 11 |
| Gene Symbols | TP53I11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.