Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TP53I13 Overexpression Lysate
SDP

Catalog No. NBL117205 Shop All R&D Systems Products
Catalog No. NBL117205 Supplier Novus Biologicals™ Supplier No. NBL117205
Add to Cart
Add to Cart
The TP53I13 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Cell Line The lysate was created in HEK293T cells, using plasmid ID RC2918 and based on accession number NM_138349. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens tumor protein p53 inducible protein 13 (TP53I13), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.