Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TP53I13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590824 Supplier Invitrogen™ Supplier No. PA590824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSWRPPGTEV TSQGPRQPSS SGAKRRRLRA ALGPQPTRSA LRFPSASPGS LKAKQSMAGI PGRESNAPSV PTVSLLPGAP GGNASSRTEA QVPNGQGSPG GCVCSSQASP APRAAAPPRA ARGPTPRTEE A; Positive Samples: Mouse liver, Mouse brain, Rat liver; Cellular Location: Cell membrane, Cytoplasm, Extracellular side, Single-pass type I membrane protein.

TP53I13 gene ontology annotations related to this gene include negative regulation of cell cycle; response to drug; response to organic cyclic compound; response to UV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TP53I13
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Trp53i13
Gene Accession No. B0BN44, Q5F267
Gene Alias 2410019G02Rik; Damage-stimulated cytoplasmic protein 1; DSCP1; LOW QUALITY PROTEIN: tumor protein p53-inducible protein 13; RGD1559818; Tp53i13; transformation related protein 53 inducible protein 13; Trp53i13; tumor protein p53 inducible protein 13; tumor protein p53-inducible protein 13
Gene Symbols TP53I13, Trp53i13
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-310 of human TP53I13 (NP_6123583).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216964, 287550
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.