Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TP53I13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569779
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
TP53I13 gene ontology annotations related to this gene include negative regulation of cell cycle; response to drug; response to organic cyclic compound; response to UV.
Specifications
TP53I13 | |
Polyclonal | |
Unconjugated | |
TP53I13 | |
2410019G02Rik; Damage-stimulated cytoplasmic protein 1; DSCP1; LOW QUALITY PROTEIN: tumor protein p53-inducible protein 13; RGD1559818; Tp53i13; transformation related protein 53 inducible protein 13; Trp53i13; tumor protein p53 inducible protein 13; tumor protein p53-inducible protein 13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
90313 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NBR0 | |
TP53I13 | |
synthetic peptide directed towards the C terminal region of human TP53I13. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction