Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TP53INP2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171704H
Description
TP53INP2 Polyclonal antibody specifically detects TP53INP2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
TP53INP2 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
C20orf110, chromosome 20 open reading frame 110, diabetes and obesity regulated, dJ1181N3.1, DKFZp434B2411, DKFZp434O0827, DOR, FLJ21759, FLJ23500, p53-inducible protein U, PINHPIG-U, tumor protein p53 inducible nuclear protein 2, tumor protein p53-inducible nuclear protein 2 | |
A genomic peptide made to a C-terminal region of the human TP53INP2 protein (within residues 100-220). [Swiss-Prot Q8IXH6] | |
0.1 mL | |
Autophagy, Cancer | |
58476 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction