Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TP53TG5 Antibody (Z38P1E4*C7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | TP53TG5 |
---|---|
Clone | Z38P1E4*C7 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
TP53TG5 Monoclonal antibody specifically detects TP53TG5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TP53TG5 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Signal Transduction | |
PBS | |
27296 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Z38P1E4*C7 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C20orf10, chromosome 20 open reading frame 10, CLG01, dJ453C12.5, TP53 target 5, TP53-inducible gene 5 protein, TP53-target gene 5 protein | |
Peptide Sequence - WRNSRVYK | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title