Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53TG5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987939
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7939 Supplier Bioss Supplier No. BS7702R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a significant role in p53/TP53-mediating signaling pathway.

Specifications

Antigen TP53TG5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TP53TG5
Gene Accession No. Q9Y2B4
Gene Alias C20orf10; CLG01; LOC100352967; P3H3; RGD1561934; TP53 target 5; TP53-inducible gene 5 protein; TP53-target gene 5 protein; TP53TG5
Gene Symbols TP53TG5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TP53TG5 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27296
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.