Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TP73-AS1 Antibody (OTI2B9), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274559C
Description
TP73-AS1 Monoclonal specifically detects TP73-AS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TP73-AS1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TP73-AS1 | |
Full length recombinant protein of human TP73-AS1 (NP_997198) | |
0.1 mL | |
57212.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2B9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
KIAA0495, MGC138189, PDAM | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction