Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TP73-AS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570359
Description
This target displays homology in the following species: Human: 100%.
TP73-AS1 in molecular biology is P73 antisense RNA 1T (non-protein coding) and also known as PDAM, is a long non-coding RNA. It may regulate apoptosis via regulation of p53-dependent anti-apoptotic genes. It may play a role in the development of oligodendroglial tumours.
Specifications
TP73-AS1 | |
Polyclonal | |
Unconjugated | |
TP73-AS1 | |
KIAA0495; PDAM; TP73 antisense RNA 1; TP73-AS1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57212 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5-1.0 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
TP73-AS1 | |
synthetic peptide directed towards the middle region of human TP73-AS1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction