Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPCN1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA558915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558915 Supplier Thermo Scientific Supplier No. PA558915

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: FKSLLLHKRT AIQHAYRLLI SQRRPAGISY RQFEGLMRFY KPRMSARERY LTFKALNQNN TPLLSLKDFY DIYEVAALKW KAKKNREHWF DELPRT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

TPCN1 may function as one of the major voltage-gated Ca2+ channel (VDCC) across the plasma membrane. Voltage-gated Ca(2+) and Na+ channels have 4 homologous domains, each containing 6 transmembrane segments, S1 to S6. TPCN1 is similar to these channels, but it has only 2 domains containing S1 to S6 (Ishibashi et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPCN1
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TPCN1
Gene Accession No. Q9ULQ1
Gene Alias 5730403B01Rik; FLJ20612; Kiaa1169; mKIAA1169; si:dkey-125i10.5; TPC1; TPCN1; Two pore calcium channel protein 1; two pore channel 1; two pore segment channel 1; two-pore channel 1; two-pore channel 1, homolog; two-pore segment channel 1; voltage-dependent calcium channel protein TPC1; voltage-gated Ca channel
Gene Symbols TPCN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TPCN1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53373
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.