Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPD52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543152
Description
Peptide sequence: AGQKASAAFS SVGSVITKKL EDVKNSPTFK SFEEKVENLK SKVGGTKPAG Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
It belongs to the TPD52 family. Its exact function remains unknown.
Specifications
| TPD52 | |
| Polyclonal | |
| Unconjugated | |
| Tpd52 | |
| 28 kDa calcium-dependent phosphoprotein; AI043038; calcium sensitive phosphoprotein (CSPP28); CSPP28; D52; hD52; LOC100058422; mD52; N8L; PC-1; pp28; PrLZ; prostate and colon associated protein; prostate leucine zipper; protein N8; si:ch211-203l9.2; si:ch211-203l9.3; TPD52; tumor promoter D52 protein; tumor protein D52; Unknown (protein for MGC:127812) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21985, 7163 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P55327, Q62393 | |
| Tpd52 | |
| Synthetic peptide directed towards the middle region of human TPD52. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction