Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TPD52L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549076
Description
TPD52L1 Polyclonal Antibody for Western Blot
This gene encodes a member of the tumor protein D52 family. The encoded protein contains a coiled-coil domain and may form homo- or hetero-dimer with TPD52 family members. The protein is reported to be involved in cell proliferation and calcium signaling. It also interacts with the mitogen-activated protein kinase kinase kinase 5 and positively regulates MAP3K5-induced apoptosis. Multiple alternatively spliced transcript variants have been observed, but the full-length nature of some variants has not yet been determined.
Specifications
TPD52L1 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
O54818, Q16890 | |
TPD52L1 | |
KLH conjugated synthetic peptide between 43-70 amino acids from the N-terminal region of human TPD52L1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TPD52L1 | |
D53; hD53; mD53; MGC8556; Tpd52l1; TPD52L2; Tumor protein D52-like 1; tumor protein D53 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
21987, 7164 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction