Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPD52L2/D54 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB271880 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271880 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271880 Supplier Novus Biologicals Supplier No. NBP236548AF532
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPD52L2/D54 Polyclonal antibody specifically detects TPD52L2/D54 in Human samples. It is validated for Western Blot

Specifications

Antigen TPD52L2/D54
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp686A1765, HCCR-binding protein 2, tumor protein D52-like 2hD54D54, tumor protein D54
Host Species Rabbit
Immunogen The immunogen recognized by the antibody maps to a region between residue 156 to 206 of human Tumor Protein D52-like 2 using the numbering given in entry NP_003279.2.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7165
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.